Assay Details
| Gene Symbol: LOC4325567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766618.2 | XP_015622104.1 | 11 - 12 | 1060 | 117 | |
| XM_015766619.2 | XP_015622105.1 | 11 - 12 | 1061 | 117 | ||
| GenBank mRNA | AK065467.1 | - | 11 - 12 | 1016 | 117 | |
| AK101456.1 | - | 11 - 12 | 1056 | 117 | ||
More Information
Set Membership: |
|

LOC4325567
Genomic Map