Assay Details
| Gene Symbol: LOC4325567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766618.2 | XP_015622104.1 | 13 - 14 | 1195 | 72 | |
| XM_015766619.2 | XP_015622105.1 | 13 - 14 | 1196 | 72 | ||
| GenBank mRNA | AK101456.1 | - | 13 - 14 | 1191 | 72 | |
More Information
Set Membership: |
|

LOC4325567
Genomic Map