Assay Details
| Gene Symbol: LOC4325567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766618.2 | XP_015622104.1 | 15 - 16 | 1395 | 91 | |
| XM_015766619.2 | XP_015622105.1 | 15 - 16 | 1396 | 91 | ||
| GenBank mRNA | AK065467.1 | - | 14 - 15 | 1549 | 91 | |
| AK101456.1 | - | 15 - 16 | 1391 | 91 | ||
More Information
Set Membership: |
|

LOC4325567
Genomic Map