Assay Details
| Gene Symbol: LOC4325567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766618.2 | XP_015622104.1 | 18 - 19 | 1719 | 142 | |
| XM_015766619.2 | XP_015622105.1 | 18 - 19 | 1720 | 142 | ||
| GenBank mRNA | AK101456.1 | - | 18 - 19 | 1715 | 142 | |
More Information
Set Membership: |
|

LOC4325567
Genomic Map