Assay Details
| Gene Symbol: LOC4325567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766618.2 | XP_015622104.1 | 23 - 24 | 2203 | 113 | |
| GenBank mRNA | AK101456.1 | - | 23 - 24 | 2199 | 113 | |
More Information
Set Membership: |
|

LOC4325567
Genomic Map