Assay Details
| Gene Symbol: LOC4327695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788843.2 | XP_015644329.1 | 1 - 2 | 285 | 80 | |
| GenBank mRNA | AK070082.1 | - | 1 - 2 | 232 | 80 | |
More Information
Set Membership: |
|

LOC4327695
Genomic Map