Assay Details
| Gene Symbol: LOC4324264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766074.2 | XP_015621560.1 | 10 - 11 | 1611 | 122 | |
| GenBank mRNA | AK072491.1 | - | 2 - 3 | 160 | 122 | |
| AK102926.1 | - | 10 - 11 | 1598 | 122 | ||
More Information
Set Membership: |
|

LOC4324264
Genomic Map