Assay Details
| Gene Symbol: LOC4324264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766074.2 | XP_015621560.1 | 12 - 13 | 1733 | 98 | |
| GenBank mRNA | AK072491.1 | - | 4 - 5 | 282 | 98 | |
| AK102926.1 | - | 12 - 13 | 1720 | 98 | ||
| AK104914.1 | - | 3 - 4 | 232 | 98 | ||
More Information
Set Membership: |
|

LOC4324264
Genomic Map