Assay Details
| Gene Symbol: LOC4324264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766074.2 | XP_015621560.1 | 14 - 15 | 1881 | 78 | |
| GenBank mRNA | AK072491.1 | - | 6 - 7 | 430 | 78 | |
| AK102926.1 | - | 14 - 15 | 1868 | 78 | ||
| AK104914.1 | - | 5 - 6 | 380 | 78 | ||
More Information
Set Membership: |
|

LOC4324264
Genomic Map