Assay Details
| Gene Symbol: LOC4324264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766074.2 | XP_015621560.1 | 15 - 16 | 1932 | 72 | |
| GenBank mRNA | AK072491.1 | - | 7 - 8 | 481 | 72 | |
| AK102926.1 | - | 15 - 16 | 1919 | 72 | ||
| AK104914.1 | - | 6 - 7 | 431 | 72 | ||
More Information
Set Membership: |
|

LOC4324264
Genomic Map