Assay Details
| Gene Symbol: LOC4324264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766074.2 | XP_015621560.1 | 2 - 3 | 366 | 120 | |
| GenBank mRNA | AK062536.1 | - | 2 - 3 | 281 | 120 | |
| AK102926.1 | - | 2 - 3 | 353 | 120 | ||
More Information
Set Membership: |
|

LOC4324264
Genomic Map