Assay Details
| Gene Symbol: LOC4324264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766074.2 | XP_015621560.1 | 7 - 8 | 1049 | 83 | |
| GenBank mRNA | AK102926.1 | - | 7 - 8 | 1036 | 83 | |
| AK108476.1 | - | 4 - 5 | 456 | 83 | ||
More Information
Set Membership: |
|

LOC4324264
Genomic Map