Assay Details
| Gene Symbol: LOC9271287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755156.2 | XP_015610642.1 | 14 - 15 | 3713 | 128 | |
| XM_015755180.2 | XP_015610666.1 | 7 - 8 | 2535 | 128 | ||
| XM_015755187.2 | XP_015610673.1 | 6 - 7 | 2433 | 128 | ||
More Information
Set Membership: |
|

LOC9271287
Genomic Map