Assay Details
| Gene Symbol: LOC9271287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755156.2 | XP_015610642.1 | 6 - 7 | 1036 | 118 | |
| XM_015755210.2 | XP_015610696.1 | 6 - 7 | 1037 | 118 | ||
| XM_015755215.2 | XP_015610701.1 | 6 - 7 | 1037 | 118 | ||
More Information
Set Membership: |
|

LOC9271287
Genomic Map