Assay Details
| Gene Symbol: LOC4327808 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766131.2 | XP_015621617.1 | 3 - 4 | 491 | 64 | |
| GenBank mRNA | AK119447.1 | - | 3 - 4 | 491 | 64 | |
More Information
Set Membership: |
|

LOC4327808
Genomic Map