Assay Details
| Gene Symbol: LOC4324792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765582.2 | 7 - 8 | 844 | 86 | ||
| XM_015765583.2 | XP_015621069.1 | 7 - 8 | 628 | 86 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4324792| Gene Symbol: LOC4324792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765582.2 | 7 - 8 | 844 | 86 | ||
| XM_015765583.2 | XP_015621069.1 | 7 - 8 | 628 | 86 | ||
Set Membership: |
|