Assay Details
| Gene Symbol: LOC4328209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771223.2 | XP_015626709.1 | 2 - 3 | 921 | 61 | |
| XM_015771224.2 | XP_015626710.1 | 2 - 3 | 921 | 61 | ||
| GenBank mRNA | AK100898.1 | - | 1 - 2 | 819 | 61 | |
More Information
Set Membership: |
|

LOC4328209
Genomic Map