Assay Details
| Gene Symbol: LOC4328209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771223.2 | XP_015626709.1 | 7 - 8 | 1346 | 93 | |
| XM_015771224.2 | XP_015626710.1 | 7 - 8 | 1346 | 93 | ||
| GenBank mRNA | AK072646.1 | - | 6 - 7 | 1246 | 93 | |
| AK100898.1 | - | 6 - 7 | 1244 | 93 | ||
More Information
Set Membership: |
|

LOC4328209
Genomic Map