Assay Details
| Gene Symbol: LOC4328239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768520.2 | XP_015624006.1 | 3 - 4 | 891 | 85 | |
| GenBank mRNA | AK102168.1 | - | 3 - 4 | 777 | 85 | |
More Information
Set Membership: |
|

LOC4328239
Genomic Map