Assay Details
| Gene Symbol: LOC4328239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768520.2 | XP_015624006.1 | 4 - 5 | 1083 | 79 | |
| GenBank mRNA | AK102168.1 | - | 4 - 5 | 969 | 79 | |
More Information
Set Membership: |
|

LOC4328239
Genomic Map