Assay Details
| Gene Symbol: LOC4328733 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770782.2 | XP_015626268.1 | 1 - 2 | 302 | 145 | |
| GenBank mRNA | AK058267.1 | - | 1 - 2 | 303 | 145 | |
More Information
Set Membership: |
|

LOC4328733
Genomic Map