Assay Details
| Gene Symbol: LOC4328733 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770782.2 | XP_015626268.1 | 7 - 8 | 1046 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4328733| Gene Symbol: LOC4328733 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770782.2 | XP_015626268.1 | 7 - 8 | 1046 | 94 | |
Set Membership: |
|