Assay Details
| Gene Symbol: LOC4328733 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770782.2 | XP_015626268.1 | 8 - 9 | 1280 | 85 | |
| GenBank mRNA | AK058267.1 | - | 8 - 9 | 1281 | 85 | |
More Information
Set Membership: |
|

LOC4328733
Genomic Map