Assay Details
Gene Symbol: LOC4347230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015755366.2 | XP_015610852.1 | 1 - 2 | 873 | 72 | |
GenBank mRNA | AK059479.1 | - | 1 - 2 | 733 | 72 | |
AK106042.1 | - | 1 - 2 | 848 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
