Assay Details
| Gene Symbol: LOC4348315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757707.2 | XP_015613193.1 | 4 - 5 | 405 | 119 | |
| GenBank mRNA | AK071372.1 | - | 4 - 5 | 401 | 119 | |
More Information
Set Membership: |
|

LOC4348315
Genomic Map