Assay Details
| Gene Symbol: LOC4350250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759824.2 | XP_015615310.1 | 5 - 6 | 899 | 68 | |
| GenBank mRNA | AK073284.1 | - | 5 - 6 | 833 | 68 | |
More Information
Set Membership: |
|

LOC4350250
Genomic Map