Assay Details
| Gene Symbol: LOC4331275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768149.2 | XP_015623635.1 | 9 - 10 | 907 | 86 | |
| XM_015768150.2 | XP_015623636.1 | 9 - 10 | 917 | 86 | ||
| GenBank mRNA | AK061331.1 | - | 8 - 9 | 888 | 86 | |
| AK067965.1 | - | 9 - 10 | 893 | 86 | ||
More Information
Set Membership: |
|

LOC4331275
Genomic Map