Assay Details
| Gene Symbol: LOC4332068 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776152.1 | XP_015631638.1 | 5 - 6 | 2460 | 68 | |
| GenBank mRNA | AK066051.1 | - | 5 - 6 | 2230 | 68 | |
| AK101307.1 | - | 2 - 3 | 306 | 68 | ||
| AK121428.1 | - | 5 - 6 | 963 | 68 | ||
More Information
Set Membership: |
|

LOC4332068
Genomic Map