Assay Details
| Gene Symbol: LOC4332333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777054.2 | XP_015632540.1 | 2 - 3 | 1086 | 72 | |
| GenBank mRNA | AK058971.1 | - | 1 - 2 | 1062 | 72 | |
More Information
Set Membership: |
|

LOC4332333
Genomic Map