Assay Details
| Gene Symbol: LOC4334356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772952.2 | XP_015628438.1 | 1 | 1245 | 68 | |
| GenBank mRNA | AK065950.1 | - | 1 | 954 | 68 | |
| DQ231252.1 | - | 1 | 912 | 68 | ||
More Information
Set Membership: |
|

LOC4334356
Genomic Map