Assay Details
| Gene Symbol: LOC4336505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777964.2 | XP_015633450.1 | 4 - 5 | 1194 | 65 | |
| XM_015777965.2 | XP_015633451.1 | 4 - 5 | 1189 | 65 | ||
| XM_015777966.1 | XP_015633452.1 | 4 - 5 | 968 | 65 | ||
| XM_015777967.2 | XP_015633453.1 | 4 - 5 | 955 | 65 | ||
| GenBank mRNA | AK066666.1 | - | 4 - 5 | 1155 | 65 | |
More Information
Set Membership: |
|

LOC4336505
Genomic Map