Assay Details
| Gene Symbol: LOC4336704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781662.2 | XP_015637148.1 | 1 - 2 | 232 | 76 | |
| GenBank mRNA | AB066661.2 | - | 1 - 2 | 230 | 76 | |
| AK069025.1 | - | 1 - 2 | 207 | 76 | ||
More Information
Set Membership: |
|

LOC4336704
Genomic Map