Assay Details
| Gene Symbol: LOC4336704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781662.2 | XP_015637148.1 | 3 - 4 | 447 | 75 | |
| GenBank mRNA | AB066661.2 | - | 3 - 4 | 445 | 75 | |
| AK069025.1 | - | 3 - 4 | 422 | 75 | ||
More Information
Set Membership: |
|

LOC4336704
Genomic Map