Assay Details
| Gene Symbol: LOC4336842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778185.2 | XP_015633671.1 | 3 - 4 | 314 | 66 | |
| XM_015778186.2 | XP_015633672.1 | 3 - 4 | 324 | 66 | ||
| XM_015778187.2 | XP_015633673.1 | 3 - 4 | 314 | 66 | ||
| XM_015778188.2 | XP_015633674.1 | 3 - 4 | 324 | 66 | ||
| XM_015778189.2 | XP_015633675.1 | 4 - 5 | 416 | 66 | ||
| XM_015778190.2 | XP_015633676.1 | 4 - 5 | 413 | 66 | ||
| GenBank mRNA | AK067019.1 | - | 3 - 4 | 296 | 66 | |
More Information
Set Membership: |
|

LOC4336842
Genomic Map