Assay Details
| Gene Symbol: LOC9268618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781419.2 | XP_015636905.1 | 6 - 7 | 788 | 147 | |
| XM_015781422.2 | XP_015636908.1 | 6 - 7 | 788 | 147 | ||
| GenBank mRNA | AK242596.1 | - | 4997 | 147 | ||
| AK288235.1 | - | 4999 | 147 | |||
More Information
Set Membership: |
|

LOC9268618
Genomic Map