Assay Details
| Gene Symbol: LOC4337050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780018.1 | XP_015635504.1 | 1 - 2 | 183 | 69 | |
| GenBank mRNA | AK071587.1 | - | 1 - 2 | 111 | 69 | |
| BT129276.1 | - | 1 - 2 | 108 | 69 | ||
| Y10118.1 | - | 1 - 2 | 70 | 69 | ||
| Gene Symbol: LOC9266366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780020.2 | XP_015635506.1 | 13 - 14 | 2719 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015780018.1, Y10118.1 transcripts.
Set Membership: |
|

LOC4337050
Genomic Map