Assay Details
| Gene Symbol: LOC4337050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780018.1 | XP_015635504.1 | 2 - 3 | 273 | 63 | |
| XM_015780019.2 | XP_015635505.1 | 2 - 3 | 229 | 63 | ||
| GenBank mRNA | AK071587.1 | - | 2 - 3 | 201 | 63 | |
| Y10118.1 | - | 2 - 3 | 160 | 63 | ||
More Information
Set Membership: |
|

LOC4337050
Genomic Map