Assay Details
| Gene Symbol: LOC4337050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780018.1 | XP_015635504.1 | 3 - 4 | 387 | 64 | |
| XM_015780019.2 | XP_015635505.1 | 3 - 4 | 343 | 64 | ||
| GenBank mRNA | AK071587.1 | - | 3 - 4 | 315 | 64 | |
| BT129276.1 | - | 2 - 3 | 405 | 64 | ||
| Y10118.1 | - | 3 - 4 | 274 | 64 | ||
| Gene Symbol: LOC9266366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780020.2 | XP_015635506.1 | 14 - 15 | 3016 | 64 | |
More Information
Set Membership: |
|

LOC4337050
Genomic Map