Assay Details
| Gene Symbol: LOC4337246 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778216.2 | XP_015633702.1 | 1 | 435 | 121 | |
| GenBank mRNA | AK062771.1 | - | 1 | 441 | 121 | |
More Information
Set Membership: |
|

LOC4337246
Genomic Map