Assay Details
| Gene Symbol: LOC4337298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780936.2 | XP_015636422.1 | 12 - 13 | 1282 | 104 | |
| XM_015780937.2 | XP_015636423.1 | 12 - 13 | 1282 | 104 | ||
| GenBank mRNA | AK102939.1 | - | 12 - 13 | 1266 | 104 | |
More Information
Set Membership: |
|

LOC4337298
Genomic Map