Assay Details
| Gene Symbol: LOC4337298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780936.2 | XP_015636422.1 | 4 - 5 | 614 | 65 | |
| XM_015780937.2 | XP_015636423.1 | 4 - 5 | 614 | 65 | ||
| GenBank mRNA | AK102939.1 | - | 4 - 5 | 598 | 65 | |
More Information
Set Membership: |
|

LOC4337298
Genomic Map