Assay Details
| Gene Symbol: LOC4338767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785265.2 | XP_015640751.1 | 1 - 2 | 733 | 65 | |
| GenBank mRNA | AK068553.1 | - | 1 - 2 | 697 | 65 | |
| AK071584.1 | - | 2 - 3 | 496 | 65 | ||
More Information
Set Membership: |
|

LOC4338767
Genomic Map