Assay Details
| Gene Symbol: LOC4338767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785265.2 | XP_015640751.1 | 6 - 7 | 1565 | 69 | |
| GenBank mRNA | AK068553.1 | - | 6 - 7 | 1529 | 69 | |
| AK071584.1 | - | 7 - 8 | 1328 | 69 | ||
More Information
Set Membership: |
|

LOC4338767
Genomic Map