Assay Details
| Gene Symbol: LOC4339175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783667.1 | XP_015639153.1 | 1 - 2 | 320 | 110 | |
| GenBank mRNA | AK061987.1 | - | 1 - 2 | 320 | 110 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015783667.1 transcripts.
Set Membership: |
|

LOC4339175
Genomic Map