Assay Details
| Gene Symbol: LOC4339175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783667.1 | XP_015639153.1 | 2 - 3 | 383 | 107 | |
| XM_015783668.2 | XP_015639154.1 | 2 - 3 | 375 | 107 | ||
| GenBank mRNA | AK061987.1 | - | 2 - 3 | 383 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015783667.1, XM_015783668.2 transcripts.
Set Membership: |
|

LOC4339175
Genomic Map