Assay Details
| Gene Symbol: LOC4339175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783666.2 | XP_015639152.1 | 2 - 3 | 517 | 72 | |
| XM_015783667.1 | XP_015639153.1 | 3 - 4 | 593 | 72 | ||
| XM_015783668.2 | XP_015639154.1 | 3 - 4 | 585 | 72 | ||
| XM_015783669.2 | XP_015639155.1 | 2 - 3 | 513 | 72 | ||
| GenBank mRNA | AK061987.1 | - | 3 - 4 | 593 | 72 | |
| AK069306.1 | - | 2 - 3 | 524 | 72 | ||
More Information
Set Membership: |
|

LOC4339175
Genomic Map