Assay Details
| Gene Symbol: LOC4339175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783666.2 | XP_015639152.1 | 3 - 4 | 604 | 87 | |
| XM_015783667.1 | XP_015639153.1 | 4 - 5 | 680 | 87 | ||
| XM_015783668.2 | XP_015639154.1 | 4 - 5 | 672 | 87 | ||
| XM_015783669.2 | XP_015639155.1 | 3 - 4 | 600 | 87 | ||
| GenBank mRNA | AK061987.1 | - | 4 - 5 | 680 | 87 | |
| AK069306.1 | - | 3 - 4 | 611 | 87 | ||
More Information
Set Membership: |
|

LOC4339175
Genomic Map