Assay Details
| Gene Symbol: LOC4339175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783666.2 | XP_015639152.1 | 5 - 6 | 708 | 77 | |
| XM_015783667.1 | XP_015639153.1 | 6 - 7 | 784 | 77 | ||
| XM_015783668.2 | XP_015639154.1 | 6 - 7 | 776 | 77 | ||
| XM_015783669.2 | XP_015639155.1 | 5 - 6 | 704 | 77 | ||
| GenBank mRNA | AK061987.1 | - | 6 - 7 | 784 | 77 | |
| AK069306.1 | - | 5 - 6 | 715 | 77 | ||
More Information
Set Membership: |
|

LOC4339175
Genomic Map