Assay Details
| Gene Symbol: LOC4341204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786038.2 | XP_015641524.1 | 3 - 4 | 1398 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4341204| Gene Symbol: LOC4341204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786038.2 | XP_015641524.1 | 3 - 4 | 1398 | 61 | |
Set Membership: |
|