Assay Details
| Gene Symbol: LOC4341387 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788295.2 | XP_015643781.1 | 1 - 2 | 646 | 83 | |
| GenBank mRNA | AF513382.1 | - | 1 - 2 | 611 | 83 | |
| AK068051.1 | - | 1 - 2 | 628 | 83 | ||
| AK068682.1 | - | 1 - 2 | 628 | 83 | ||
More Information
Set Membership: |
|

LOC4341387
Genomic Map